Gene Transcripts

Gene Symbol
Krtap31-1
Entrez Gene ID
70831
Gene Name
keratin associated protein 31-1
Gene Aliases
4733401H21Rik, AI506529
Chromosome Location
Chr. 11: 99907920 - 99908890 on Build GRCm38
UniGene ID
Mm.347981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027568.2 NP_081844.1 1 774
GenBank mRNA AK014635.1 - 1 775

View Full Product Details