Gene Transcripts

Gene Symbol
Kcnip1
Entrez Gene ID
70357
Gene Name
Kv channel-interacting protein 1
Gene Aliases
KCHIP1, Kchip1.2
Chromosome Location
Chr. 11: 33629339 - 33993193 on Build GRCm38
UniGene ID
Mm.252514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190885.1 NP_001177814.1 9 1636
NM_001190886.1 NP_001177815.1 8 1592
NM_001290690.1 NP_001277619.1 8 1791
NM_027398.3 NP_081674.2 8 1603
GenBank mRNA AK014313.1 - 8 1591
AK045948.1 - 9 1637
AK046398.1 - 9 1625
AK081845.1 - 8 1593
AY171234.1 - 8 1665
BC034241.1 - 8 1545 4503251
DQ148493.1 - 8 1545
DQ148494.1 - 8 1593

View Full Product Details