Gene Transcripts

Gene Symbol
Slc39a4
Entrez Gene ID
72027
Gene Name
solute carrier family 39 (zinc transporter), member 4
Gene Aliases
1600025H15Rik, AU041686, AWMS2, ZIP4
Chromosome Location
Chr. 15: 76612383 - 76617379 on Build GRCm38
UniGene ID
Mm.276829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028064.2 NP_082340.1 5 1083
GenBank mRNA AK005535.1 - 5 1123
AK146977.1 - 5 1124
AK147107.1 - 5 1102
AK154089.1 - 5 1602
AK155467.1 - 5 1602
BC023498.1 - 5 1083 5011088

View Full Product Details