Gene Transcripts

Gene Symbol
Trub1
Entrez Gene ID
72133
Gene Name
TruB pseudouridine (psi) synthase homolog 1 (E. coli)
Gene Aliases
2610009I02Rik, 9030425C13Rik, AI448235, BB118943, C76870
Chromosome Location
Chr. 19: 57452891 - 57491005 on Build GRCm38
UniGene ID
Mm.258404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028115.3 NP_082391.1 2 333
NM_029839.3 NP_084115.2 2 333
GenBank mRNA AK011362.1 - 2 328
AK018525.1 - 2 306
AK031674.1 - 2 311
AK164095.1 - 2 304
AK169221.1 - 2 299
BC039287.1 - 2 278
BC054088.1 - 2 147
BC089592.1 - 2 300 6757213
BY094716.1 - 2 335

View Full Product Details