Gene Transcripts
- Gene Symbol
- Tars2
- Entrez Gene ID
- 71807
- Gene Name
- threonyl-tRNA synthetase 2, mitochondrial (putative)
- Gene Aliases
- 2610024N01Rik, AI429208, Tarsl1
- Chromosome Location
- Chr. 3: 95739974 - 95754977 on Build GRCm38
- UniGene ID
- Mm.271754
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163617.1 |
NP_001157089.1
|
5, 6 |
792 |
|
NM_001163619.1 |
NP_001157091.1
|
5, 6 |
792 |
|
NM_027931.3 |
NP_082207.2
|
5, 6 |
792 |
|
GenBank mRNA |
AK011540.1 |
- |
5, 6 |
678 |
|
AK080690.1 |
- |
5, 6 |
697 |
|
AK088675.1 |
- |
4, 5 |
542 |
|
AK142685.1 |
- |
5, 6 |
687 |
|
AK164589.1 |
- |
5, 6 |
699 |
|
AK164825.1 |
- |
5, 6 |
650 |
|
AK199920.1 |
- |
1, 2 |
53 |
|
BC008995.1 |
- |
5, 6 |
677 |
3157108 |
BC147501.1 |
- |
5, 6 |
676 |
9056383 |
View Full Product Details