Gene Transcripts

Gene Symbol
Igsf5
Entrez Gene ID
72058
Gene Name
immunoglobulin superfamily, member 5
Gene Aliases
2010003D20Rik, JCAM, Jam4
Chromosome Location
Chr. 16: 96360467 - 96465572 on Build GRCm38
UniGene ID
Mm.119714
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177887.1 NP_001171358.1 9 1205
NM_028078.3 NP_082354.1 8 898
GenBank mRNA AF537215.1 - 9 1139
AK008060.1 - 8 869
AK078934.1 - 8 869
BC004806.1 - 8 851 3584645

View Full Product Details