Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000260.3 | NP_000251.3 | 33 | 4695 | |
NM_001127180.1 | NP_001120652.1 | 33 | 4689 | ||
XM_011545044.2 | XP_011543346.1 | 33 | 4767 | ||
XM_011545046.2 | XP_011543348.2 | 32 | 4528 | ||
XM_011545050.2 | XP_011543352.1 | 31 | 4490 | ||
XM_017017778.1 | XP_016873267.1 | 33 | 4636 | ||
XM_017017779.1 | XP_016873268.1 | 33 | 4636 | ||
XM_017017780.1 | XP_016873269.1 | 32 | 4528 | ||
XM_017017781.1 | XP_016873270.1 | 32 | 4546 | ||
XM_017017782.1 | XP_016873271.1 | 32 | 4528 | ||
XM_017017783.1 | XP_016873272.1 | 32 | 4528 | ||
XM_017017784.1 | XP_016873273.1 | 32 | 4528 | ||
XM_017017785.1 | XP_016873274.1 | 31 | 4405 | ||
XM_017017786.1 | XP_016873275.1 | 32 | 4528 | ||
XM_017017788.1 | XP_016873277.1 | 32 | 4528 | ||
GenBank mRNA | AB209629.1 | - | 14 | 2076 | |
AB290181.1 | - | 13 | 1966 | ||
BC144519.1 | - | 34 | 4737 | ||
U39226.1 | - | 33 | 4695 | ||
U55208.1 | - | 33 | 4690 |