Gene Transcripts
- Gene Symbol
- Nfam1
- Entrez Gene ID
- 74039
- Gene Name
- Nfat activating molecule with ITAM motif 1
- Gene Aliases
- 4921501M20Rik, Cnaip
- Chromosome Location
- Chr. 15: 82996736 - 83033397 on Build GRCm38
- UniGene ID
- Mm.32584
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271411.1 |
NP_001258340.1
|
5, 6 |
910 |
|
NM_001271412.1 |
NP_001258341.1
|
5, 6 |
840 |
|
NM_001271413.1 |
NP_001258342.1
|
5, 6 |
749 |
|
NM_001271414.1 |
NP_001258343.1
|
5, 6 |
837 |
|
NM_028728.3 |
NP_083004.1
|
5, 6 |
767 |
|
GenBank mRNA |
AF361364.1 |
- |
5, 6 |
844 |
|
AK014802.2 |
- |
5, 6 |
768 |
|
AK048992.1 |
- |
5, 6 |
751 |
|
AK150635.1 |
- |
5, 6 |
819 |
|
AK151187.1 |
- |
5, 6 |
819 |
|
AK151884.1 |
- |
5, 6 |
819 |
|
AK152262.1 |
- |
5, 6 |
819 |
|
AK152499.1 |
- |
5, 6 |
819 |
|
AK152688.1 |
- |
5, 6 |
819 |
|
AK155106.1 |
- |
5, 6 |
764 |
|
AK162939.1 |
- |
5, 6 |
821 |
|
AK169918.1 |
- |
5, 6 |
764 |
|
AK170098.1 |
- |
5, 6 |
854 |
|
AK170355.1 |
- |
5, 6 |
817 |
|
AK170442.1 |
- |
5, 6 |
819 |
|
AK170545.1 |
- |
5, 6 |
705 |
|
AK170593.1 |
- |
5, 6 |
775 |
|
AK170677.1 |
- |
5, 6 |
775 |
|
AK170811.1 |
- |
5, 6 |
775 |
|
AK170883.1 |
- |
5, 6 |
818 |
|
AK171082.1 |
- |
5, 6 |
880 |
|
AK171086.1 |
- |
5, 6 |
788 |
|
AK171201.1 |
- |
5, 6 |
749 |
|
BC150153.1 |
- |
5, 6 |
782 |
|
BC150488.1 |
- |
5, 6 |
782 |
40141021 |
View Full Product Details