Gene Transcripts

Gene Symbol
Scara5
Entrez Gene ID
71145
Gene Name
scavenger receptor class A, member 5 (putative)
Gene Aliases
4932433F15Rik, 4933425F03Rik, AV278087, Tesr
Chromosome Location
Chr. 14: 65666403 - 65764826 on Build GRCm38
UniGene ID
Mm.83840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168318.1 NP_001161790.1 2, 3 466
NM_028903.2 NP_083179.2 2, 3 466
GenBank mRNA AK016912.1 - 2, 3 433
AK033154.1 - 2, 3 433
AK133176.1 - 2, 3 467
BC016096.1 - 2, 3 442 4506472
BC023907.1 - 2, 3 382 5321978
BC032159.1 - 2, 3 422 5354657
DQ122126.1 - 1, 2 116

View Full Product Details