Gene Transcripts
- Gene Symbol
- Lrrcc1
- Entrez Gene ID
- 71710
- Gene Name
- leucine rich repeat and coiled-coil domain containing 1
- Gene Aliases
- 1200008A14Rik, 4932441F23Rik, AI195358, AI447421
- Chromosome Location
- Chr. 3: 14533788 - 14572658 on Build GRCm38
- UniGene ID
- Mm.257449
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163579.1 |
NP_001157051.1
|
1, 2 |
233 |
|
NM_001163580.1 |
NP_001157052.1
|
1, 2 |
233 |
|
NM_028915.3 |
NP_083191.2
|
1, 2 |
233 |
|
GenBank mRNA |
AK004646.1 |
- |
1, 2 |
234 |
|
AK029563.1 |
- |
1, 2 |
150 |
|
AK173256.1 |
- |
1, 2 |
161 |
|
BC061202.1 |
- |
1, 2 |
189 |
|
BC099680.1 |
- |
1, 2 |
197 |
|
BC138734.1 |
- |
1, 2 |
207 |
8861756 |
BC138735.1 |
- |
1, 2 |
207 |
8861757 |
View Full Product Details