Gene Transcripts

Gene Symbol
Lmbr1l
Entrez Gene ID
74775
Gene Name
limb region 1 like
Gene Aliases
1110013E13Rik, AA415653, D15Ertd735e, Limr
Chromosome Location
Chr. 15: 98903914 - 98918550 on Build GRCm38
UniGene ID
Mm.385020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029098.3 NP_083374.1 5 600
GenBank mRNA AK003656.1 - 5 532
AK030589.1 - 5 566
AK049110.1 - 5 601
AK143891.1 - 5 561
AK154968.1 - 5 734
AK170328.1 - 5 561
AK173121.1 - 5 981
AY052398.1 - 5 443
BC023397.1 - 5 537 5043877

View Full Product Details