Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029770.2 | NP_084046.2 | 4 | 913 | |
GenBank mRNA | AJ487853.1 | - | 4 | 816 | |
AK018177.1 | - | 4 | 913 | ||
AK147603.1 | - | 4 | 808 | ||
AK154271.1 | - | 4 | 913 | ||
AK154287.1 | - | 4 | 916 | ||
AK158284.1 | - | 4 | 913 | ||
AK170513.1 | - | 4 | 912 | ||
BC048162.1 | - | 4 | 720 | ||
BC057560.1 | - | 4 | 720 | 6417563 |