Gene Transcripts

Gene Symbol
Cog2
Entrez Gene ID
76332
Gene Name
component of oligomeric golgi complex 2
Gene Aliases
1190002B08Rik, 2700012E02Rik, C86089, Ldlc
Chromosome Location
Chr. 8: 124520767 - 124552008 on Build GRCm38
UniGene ID
Mm.240108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029746.3 NP_084022.2 13 1537
GenBank mRNA AK004441.1 - 1 15
AK089920.1 - 13 1521
AK157348.1 - 13 1499
AK169298.1 - 13 1507
AK171726.1 - 13 1456
AK187156.1 - 3 225
BC011435.1 - 13 1477 4010102
W74920.1 - 4 422

View Full Product Details