Gene Transcripts
- Gene Symbol
- Cttnbp2nl
- Entrez Gene ID
- 80281
- Gene Name
- CTTNBP2 N-terminal like
- Gene Aliases
- AA552995, AA589392, AU018624, BC003236, mKIAA1433
- Chromosome Location
- Chr. 3: 105001915 - 105053146 on Build GRCm38
- UniGene ID
- Mm.200327
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163332.1 |
NP_001156804.1
|
3, 4 |
293 |
|
NM_001163333.1 |
NP_001156805.1
|
3, 4 |
285 |
|
NM_030249.4 |
NP_084525.1
|
3, 4 |
401 |
|
GenBank mRNA |
AK029901.1 |
- |
3, 4 |
402 |
|
AK030438.1 |
- |
3, 4 |
266 |
|
AK051259.1 |
- |
3, 4 |
265 |
|
AK129359.1 |
- |
3, 4 |
291 |
|
AK145085.1 |
- |
3, 4 |
264 |
|
BC003236.1 |
- |
3, 4 |
255 |
3498078 |
BY195820.1 |
- |
3, 4 |
298 |
|
CV562515.1 |
- |
3, 4 |
285 |
|
View Full Product Details