Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030731.3 | NP_109656.1 | 3 | 2449 | |
GenBank mRNA | AK029533.1 | - | 4 | 597 | |
AK030843.1 | - | 4 | 597 | ||
AK030862.1 | - | 3 | 2450 | ||
AK036210.1 | - | 4 | 598 | ||
AK039280.1 | - | 4 | 609 | ||
AK042070.1 | - | 4 | 600 | ||
AK088922.1 | - | 4 | 597 | ||
BC056390.1 | - | 4 | 590 | 6854944 | |
BC059017.1 | - | 4 | 590 | 6848690 |