Gene Transcripts
- Gene Symbol
- PON1
- Entrez Gene ID
- 5444
- Gene Name
- paraoxonase 1
- Gene Aliases
- ESA, MVCD5, PON
- Chromosome Location
- Chr. 7: 95298357 - 95324572 on Build GRCh38
- UniGene ID
- Hs.370995
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000446.5 |
NP_000437.3
|
2, 3 |
244 |
|
GenBank mRNA |
AK314027.1 |
- |
2, 3 |
170 |
|
BC074719.2 |
- |
2, 3 |
176 |
30915169 |
BJ993711.1 |
- |
2, 3 |
244 |
|
CB122285.1 |
- |
2, 3 |
204 |
|
D84371.1 |
- |
2, 3 |
201 |
|
M63012.1 |
- |
2, 3 |
156 |
|
M63013.1 |
- |
2, 3 |
95 |
|
M63014.1 |
- |
1, 2 |
61 |
|
S64615.1 |
- |
2, 3 |
156 |
|
U53784.1 |
- |
2, 3 |
201 |
|
Z70723.1 |
- |
2, 3 |
201 |
|
View Full Product Details