Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083322.2 | NP_001076791.1 | 3 | 182 | |
NM_001286018.1 | NP_001272947.1 | 3 | 182 | ||
NM_033078.4 | NP_149069.1 | 3 | 275 | ||
GenBank mRNA | AA174479.1 | - | 3 | 291 | |
AF030313.1 | - | 3 | 159 | ||
AF039026.1 | - | 3 | 189 | ||
AF054819.1 | - | 4 | 449 | ||
AK040436.1 | - | 3 | 264 | ||
AK170499.1 | - | 3 | 183 | ||
AY860972.1 | - | 3 | 165 | ||
BC057147.1 | - | 3 | 226 | 5328432 | |
CT010384.1 | - | 3 | 165 |