Gene Transcripts

Gene Symbol
TPMT
Entrez Gene ID
7172
Gene Name
thiopurine S-methyltransferase
Gene Aliases
TPMTD
Chromosome Location
Chr. 6: 18128311 - 18155165 on Build GRCh38
UniGene ID
Hs.444319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000367.3 NP_000358.1 9 842
XM_011514839.2 XP_011513141.1 8 755
XM_017011241.1 XP_016866730.1 10 1136
GenBank mRNA AF021876.1 - 8 665
AF021877.1 - 8 665
BC005339.1 - 9 752 3829338
BC009596.1 - 9 751 4133029
S62904.1 - 8 730
U12387.1 - 9 741

View Full Product Details