Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033523.4 | NP_277058.1 | 3 | 773 | |
GenBank mRNA | AB063496.1 | - | 3 | 284 | |
AK036430.1 | - | 3 | 774 | ||
AK153700.1 | - | 3 | 801 | ||
AK157817.1 | - | 3 | 775 | ||
BC066013.1 | - | 3 | 843 | 5706237 |