Gene Transcripts

Gene Symbol
Tssk6
Entrez Gene ID
83984
Gene Name
testis-specific serine kinase 6
Gene Aliases
Sstk
Chromosome Location
Chr. 8: 69902215 - 69903518 on Build GRCm38
UniGene ID
Mm.432504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032004.1 NP_114393.1 1 1176
GenBank mRNA AF329484.1 - 1 1176
AK076653.1 - 1 903
BC049542.1 - 1 1207

View Full Product Details