Gene Transcripts

Gene Symbol
Rpl10
Entrez Gene ID
110954
Gene Name
ribosomal protein L10
Gene Aliases
D0HXS648, DXHXS648, DXHXS648E, QM, QM16
Chromosome Location
Chr. X: 74270816 - 74273135 on Build GRCm38
UniGene ID
Mm.100113
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052835.3 NP_443067.1 5 336
GenBank mRNA AK011022.1 - 5 296
AK012561.1 - 4 293
AK014054.1 - 5 298
AK088777.1 - 5 297
AK144831.1 - 5 296
AK146217.1 - 5 297
AK151416.1 - 5 296
AK151655.1 - 5 296
AK151736.1 - 5 295
AK152123.1 - 5 296
AK152854.1 - 5 296
AK153345.1 - 5 296
AK160618.1 - 5 296
AK166610.1 - 5 295
AK167239.1 - 4 332
AK167966.1 - 5 296
AK168250.1 - 5 295
AK168854.1 - 5 296
AK187587.1 - 3 189
AK210081.1 - 3 185
BC006068.1 - 4 255
BC024901.1 - 4 257 4224016
BC048872.1 - 4 260 5052784
BC082293.1 - 4 257 6486491
BC083327.1 - 4 257 5684015
BC086917.1 - 5 295 6768088
BC092383.1 - 4 281 5710462
BC098204.1 - 4 281 6410755
BC099437.1 - 4 260 30843551
M93980.1 - 5 295
U89405.1 - 2 165
X75312.1 - 4 288

View Full Product Details