Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052994.2 | NP_443720.1 | 3, 4 | 454 | |
GenBank mRNA | AJ278999.1 | - | 3, 4 | 456 | |
AK122233.1 | - | 3, 4 | 116 | ||
AK140483.1 | - | 3, 4 | 360 | ||
AK162941.1 | - | 3, 4 | 359 | ||
BC057324.1 | - | 3, 4 | 454 | 6821449 | |
BC060210.1 | - | 3, 4 | 406 | 6838067 |