Gene Transcripts
- Gene Symbol
- Rbfox2
- Entrez Gene ID
- 93686
- Gene Name
- RNA binding protein, fox-1 homolog (C. elegans) 2
- Gene Aliases
- Fbm2, Fxh, Hrnbp2, Rbm9
- Chromosome Location
- Chr. 15: 77078990 - 77307053 on Build GRCm38
- UniGene ID
- Mm.202774
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110827.2 |
NP_001104297.1
|
5, 6 |
1175 |
|
NM_001110828.2 |
NP_001104298.1
|
4, 5 |
717 |
|
NM_001110829.2 |
NP_001104299.1
|
4, 5 |
717 |
|
NM_001110830.2 |
NP_001104300.1
|
4, 5 |
717 |
|
NM_001286417.1 |
NP_001273346.1
|
4, 5 |
717 |
|
NM_053104.6 |
NP_444334.3
|
5, 6 |
1175 |
|
NM_175387.3 |
NP_780596.1
|
5, 6 |
1175 |
|
GenBank mRNA |
AF229055.1 |
- |
4, 5 |
660 |
|
AF387322.1 |
- |
4, 5 |
461 |
|
AK044929.1 |
- |
4, 5 |
692 |
|
AK077594.1 |
- |
5, 6 |
1051 |
|
AK139555.1 |
- |
6, 7 |
1145 |
|
AY659951.1 |
- |
4, 5 |
461 |
|
AY659952.1 |
- |
4, 5 |
461 |
|
AY904025.1 |
- |
6, 7 |
611 |
|
BC002124.1 |
- |
4, 5 |
555 |
3494630 |
BC027263.1 |
- |
5, 6 |
727 |
3661748 |
DQ017388.1 |
- |
4, 5 |
461 |
|
View Full Product Details