Gene Transcripts

Gene Symbol
Optc
Entrez Gene ID
269120
Gene Name
opticin
Gene Aliases
Chromosome Location
Chr. 1: 133897193 - 133909714 on Build GRCm38
UniGene ID
Mm.59985
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160420.2 NP_001153892.1 3 363
NM_001160421.2 NP_001153893.1 2 474
NM_001160422.1 NP_001153894.1 2 474
NM_054076.3 NP_473417.2 2 474
GenBank mRNA AA223016.1 - 2 474
AF333980.1 - 2 473
AK053318.1 - 2 257
AK153750.1 - 3 364
AY077682.1 - 2 466
BC099435.1 - 2 307 30788813
BC115928.1 - 2 427 40104738

View Full Product Details