Gene Transcripts

Gene Symbol
Pip4k2c
Entrez Gene ID
117150
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, gamma
Gene Aliases
Pip5k2c
Chromosome Location
Chr. 10: 127197067 - 127211644 on Build GRCm38
UniGene ID
Mm.22682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054097.3 NP_473438.1 6, 7 812
GenBank mRNA AB054591.1 - 6, 7 755
AK148299.1 - 6, 7 772
AK208052.1 - 3, 4 225
BC005539.1 - 1, 2 26
BC021383.1 - 6, 7 745 5042933

View Full Product Details