Gene Transcripts

Gene Symbol
Vps4a
Entrez Gene ID
116733
Gene Name
vacuolar protein sorting 4a (yeast)
Gene Aliases
4930589C15Rik, AI325971, AW553189
Chromosome Location
Chr. 8: 107031326 - 107045756 on Build GRCm38
UniGene ID
Mm.236004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_126165.1 NP_569053.1 3 312
GenBank mRNA AK047821.1 - 3 324
AK159117.1 - 3 338
AK169949.1 - 3 300
BC018368.1 - 3 312 3963907

View Full Product Details