Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133245.1 | NP_573508.1 | 270 | ||
GenBank mRNA | AF060220.1 | - | 254 | ||
AF364516.1 | - | 253 | |||
AK011084.1 | - | 270 | |||
AK028172.1 | - | 270 | |||
AK146724.1 | - | 264 | |||
BC110627.2 | - | 248 | 40051901 |