Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285920.1 | NP_001272849.1 | 20 | 3965 | |
NM_001285921.1 | NP_001272850.1 | 20 | 3948 | ||
NM_001285922.1 | NP_001272851.1 | 19 | 3879 | ||
NM_001285923.1 | NP_001272852.1 | 19 | 3879 | ||
NM_133201.3 | NP_573464.2 | 19 | 3882 | ||
GenBank mRNA | AF384100.1 | - | 20 | 3761 | |
AK042080.1 | - | 19 | 3663 | ||
AK052689.1 | - | 19 | 3664 | ||
AK122220.1 | - | 18 | 4857 | ||
AK146801.1 | - | 20 | 3725 | ||
AK147863.1 | - | 19 | 3678 | ||
AK169373.1 | - | 19 | 3667 | ||
AK187724.1 | - | 1 | 232 | ||
AK211774.1 | - | 1 | 174 | ||
AK213380.1 | - | 1 | 148 | ||
AK213899.1 | - | 1 | 168 | ||
AK214723.1 | - | 1 | 231 | ||
AY123975.1 | - | 19 | 3828 | ||
BC046503.1 | - | 19 | 3660 | 4191179 |