Gene Transcripts
- Gene Symbol
- Dnah5
- Entrez Gene ID
- 110082
- Gene Name
- dynein, axonemal, heavy chain 5
- Gene Aliases
- AU022615, Dnahc5, Mdnah5, b2b1134Clo, b2b1154Clo, b2b1537Clo, b2b1565Clo, b2b601Clo, mKIAA1603
- Chromosome Location
- Chr. 15: 28154854 - 28472045 on Build GRCm38
- UniGene ID
- Mm.248464
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133365.3 |
NP_579943.3
|
33, 34 |
5696 |
|
GenBank mRNA |
AF466704.1 |
- |
33, 34 |
5697 |
|
View Full Product Details