Gene Transcripts
- Gene Symbol
- Stk17b
- Entrez Gene ID
- 98267
- Gene Name
- serine/threonine kinase 17b (apoptosis-inducing)
- Gene Aliases
- 3110009A03Rik, AI120141, Drak2
- Chromosome Location
- Chr. 1: 53755512 - 53785250 on Build GRCm38
- UniGene ID
- Mm.25559
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133810.3 |
NP_598571.2
|
2, 3 |
315 |
|
GenBank mRNA |
AK029551.1 |
- |
2, 3 |
255 |
|
AK035693.1 |
- |
2, 3 |
283 |
|
AK076932.1 |
- |
2, 3 |
282 |
|
AK083332.1 |
- |
2, 3 |
284 |
|
AK153108.1 |
- |
2, 3 |
259 |
|
AK165291.1 |
- |
2, 3 |
255 |
|
AY092028.1 |
- |
2, 3 |
268 |
|
BC006579.1 |
- |
2, 3 |
270 |
3586767 |
BY034689.1 |
- |
2, 3 |
325 |
|
View Full Product Details