Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000493.3 | NP_000484.2 | 3 | 2585 | |
XM_006715333.3 | XP_006715396.1 | 3 | 3304 | ||
XM_011535432.2 | XP_011533734.1 | 3 | 4232 | ||
XM_011535433.2 | XP_011533735.1 | 4 | 4379 | ||
XM_017010248.1 | XP_016865737.1 | 2 | 6032 | ||
XM_017010249.1 | XP_016865738.1 | 3 | 7710 |