Gene Transcripts

Gene Symbol
Spg21
Entrez Gene ID
27965
Gene Name
spastic paraplegia 21 homolog (human)
Gene Aliases
ACP33, BM-019, C78576, D9Wsu18e, GL010, MAST
Chromosome Location
Chr. 9: 65460937 - 65488470 on Build GRCm38
UniGene ID
Mm.272475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138584.2 NP_613050.1 3 378
GenBank mRNA AK009332.1 - 3 370
AK011985.1 - 3 345
AK090017.1 - 3 366
AK149930.1 - 3 352
AK150215.1 - 3 379
AK150581.1 - 3 356
AK153284.1 - 3 340
AK170666.1 - 3 371
BC099434.1 - 2 143 30714184
BC132377.1 - 3 355 40130654
BC132403.1 - 3 355 40130680

View Full Product Details