Gene Transcripts

Gene Symbol
Pim2
Entrez Gene ID
18715
Gene Name
proviral integration site 2
Gene Aliases
DXCch3, Pim-2
Chromosome Location
Chr. X: 7878306 - 7883432 on Build GRCm38
UniGene ID
Mm.347478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138606.2 NP_613072.1 4 567
GenBank mRNA BC022727.1 - 1 108
BC027376.1 - 4 487 4954711
L41495.1 - 4 567

View Full Product Details