Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301094.1 | NP_001288023.1 | 5 | 2293 | |
NM_001301096.1 | NP_001288025.1 | 4 | 2191 | ||
NM_138648.2 | NP_619589.2 | 8 | 2815 | ||
GenBank mRNA | AF303744.1 | - | 8 | 2985 | |
AK154687.1 | - | 8 | 2816 | ||
AK160919.1 | - | 8 | 2816 |