Gene Transcripts
- Gene Symbol
- Pkhd1l1
- Entrez Gene ID
- 192190
- Gene Name
- polycystic kidney and hepatic disease 1-like 1
- Gene Aliases
- AB055648, PKHDL1
- Chromosome Location
- Chr. 15: 44457371 - 44601376 on Build GRCm38
- UniGene ID
- Mm.211494
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138674.2 |
NP_619615.2
|
39 |
5790 |
|
GenBank mRNA |
AY219182.1 |
- |
39 |
5790 |
|
View Full Product Details