Gene Transcripts

Gene Symbol
Spag4
Entrez Gene ID
245865
Gene Name
sperm associated antigen 4
Gene Aliases
1700041K21Rik, MNCb-0953, Sun4, mKIAA4118
Chromosome Location
Chr. 2: 156065015 - 156069501 on Build GRCm38
UniGene ID
Mm.330713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK043812.1 - 9 1928

View Full Product Details