Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144512.2 | NP_653095.1 | 2 | 122 | |
GenBank mRNA | AK002794.1 | - | 2 | 129 | |
AK078399.1 | - | 2 | 129 | ||
AK149557.1 | - | 2 | 130 | ||
BC023117.1 | - | 2 | 110 | 4240641 | |
BC029637.1 | - | 2 | 122 | 4195373 |