Gene Transcripts
- Gene Symbol
- Slc30a6
- Entrez Gene ID
- 210148
- Gene Name
- solute carrier family 30 (zinc transporter), member 6
- Gene Aliases
- 9530029F08Rik, ZnT6
- Chromosome Location
- Chr. 17: 74395608 - 74424229 on Build GRCm38
- UniGene ID
- Mm.243943
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252478.1 |
NP_001239407.1
|
2, 3 |
155 |
|
NM_144798.6 |
NP_659047.2
|
2, 3 |
155 |
|
GenBank mRNA |
AF233346.1 |
- |
1, 2 |
107 |
|
AF395840.1 |
- |
1, 2 |
97 |
|
AK082907.1 |
- |
2, 3 |
132 |
|
AK150342.1 |
- |
2, 3 |
131 |
|
AK151120.1 |
- |
2, 3 |
131 |
|
AK160903.1 |
- |
2, 3 |
144 |
|
BC066162.1 |
- |
2, 3 |
122 |
30471027 |
BC068169.1 |
- |
2, 3 |
144 |
30542277 |
View Full Product Details