Gene Transcripts
- Gene Symbol
- Catsper1
- Entrez Gene ID
- 225865
- Gene Name
- cation channel, sperm associated 1
- Gene Aliases
- Catsper, KSper
- Chromosome Location
- Chr. 19: 5335602 - 5344281 on Build GRCm38
- UniGene ID
- Mm.87321
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139301.2 |
NP_647462.1
|
2, 3 |
1146 |
|
GenBank mRNA |
AF407332.1 |
- |
2, 3 |
1146 |
|
View Full Product Details