Gene Transcripts

Gene Symbol
Zfp410
Entrez Gene ID
52708
Gene Name
zinc finger protein 410
Gene Aliases
Apa1, D12Ertd748e, Znf410
Chromosome Location
Chr. 12: 84315928 - 84344439 on Build GRCm38
UniGene ID
Mm.255150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252582.1 NP_001239511.1 4 707
NM_001252583.1 NP_001239512.1 4 672
NM_144833.3 NP_659082.1 4 672
GenBank mRNA AF295806.1 - 4 434
AK033090.1 - 4 449
AK042224.1 - 4 429
AK049384.1 - 5 727
AK051939.1 - 2 71
AK053001.1 - 4 397
AK083003.1 - 4 414
AK132345.1 - 4 410
AK139797.1 - 3 226
BC021528.1 - 4 427 5353673
BC052080.1 - 4 404 6403411

View Full Product Details