Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145380.2 | NP_663355.1 | 2 | 154 | |
GenBank mRNA | AK077480.1 | - | 2 | 155 | |
AK084738.1 | - | 2 | 173 | ||
AK146222.1 | - | 2 | 186 | ||
AK168066.1 | - | 2 | 171 | ||
BC005598.1 | - | 2 | 154 | 3969335 | |
BC103795.1 | - | 2 | 154 | 30135815 | |
BC116787.1 | - | 2 | 151 | 40126106 | |
BC116789.1 | - | 2 | 151 | 40126108 |