Gene Transcripts
- Gene Symbol
- Ythdf2
- Entrez Gene ID
- 213541
- Gene Name
- YTH domain family 2
- Gene Aliases
- 9430020E02Rik, HGRG8, NY-REN-2
- Chromosome Location
- Chr. 4: 132184916 - 132212256 on Build GRCm38
- UniGene ID
- Mm.23998
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145393.4 |
NP_663368.3
|
3, 4 |
461 |
|
GenBank mRNA |
AK031616.1 |
- |
3, 4 |
332 |
|
AK034655.1 |
- |
3, 4 |
462 |
|
AK083882.1 |
- |
3, 4 |
260 |
|
AK144209.1 |
- |
3, 4 |
461 |
|
AK150350.1 |
- |
3, 4 |
315 |
|
AK150669.1 |
- |
3, 4 |
315 |
|
AK151392.1 |
- |
3, 4 |
315 |
|
AK151862.1 |
- |
3, 4 |
315 |
|
AK159548.1 |
- |
3, 4 |
318 |
|
AK188854.1 |
- |
3, 4 |
184 |
|
BC014797.1 |
- |
3, 4 |
267 |
3962417 |
BC028994.1 |
- |
3, 4 |
262 |
5355331 |
View Full Product Details