Gene Transcripts

Gene Symbol
BC003331
Entrez Gene ID
226499
Gene Name
cDNA sequence BC003331
Gene Aliases
1810053E15Rik, Odr4, odr-4
Chromosome Location
Chr. 1: 150361311 - 150393055 on Build GRCm38
UniGene ID
Mm.222093
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145511.2 NP_663486.2 8 1078
GenBank mRNA BC003331.1 - 7 733
CX243768.1 - 4 326
DQ077804.2 - 7 692

View Full Product Details