Gene Transcripts

Gene Symbol
Rasgef1b
Entrez Gene ID
320292
Gene Name
RasGEF domain family, member 1B
Gene Aliases
4732452O09Rik, GPIgamma4, Gpig4
Chromosome Location
Chr. 5: 99217420 - 99252927 on Build GRCm38
UniGene ID
Mm.259074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145839.2 NP_665838.1 3 474
NM_181318.4 NP_851835.1 3 474
GenBank mRNA AK028756.1 - 3 366
AK090085.1 - 3 366
AK149812.1 - 3 393
AY129963.1 - 3 401
BC050858.1 - 3 407 6517502
BC052726.1 - 3 365 6813357

View Full Product Details