Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013276.2 | NP_037408.2 | 2, 3 | 421 | |
GenBank mRNA | AF163573.1 | - | 2, 3 | 421 | |
AK055609.1 | - | 2, 3 | 347 | ||
AK312428.1 | - | 2, 3 | 368 | ||
AL832420.1 | - | 2, 3 | 360 | ||
BC020543.1 | - | 2, 3 | 377 | 4413491 |