Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
Chromosome Location
Chr. 3: 159988836 - 159996019 on Build GRCh38
UniGene ID
Hs.673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000882.3 NP_000873.2 2, 3 493
GenBank mRNA AF101062.1 - 2, 3 272
AF180562.1 - 2, 3 331
BC104982.1 - 2, 3 425 8143985
BC104984.1 - 2, 3 425 8143987
BG702253.1 - 2, 3 586
M65271.1 - 2, 3 339
M65291.1 - 2, 3 373

View Full Product Details