Gene Transcripts
- Gene Symbol
- ADH1C
- Entrez Gene ID
- 126
- Gene Name
- alcohol dehydrogenase 1C (class I), gamma polypeptide
- Gene Aliases
- ADH3
- Chromosome Location
- Chr. 4: 99336492 - 99353045 on Build GRCh38
- UniGene ID
- Hs.654537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000669.4 |
NP_000660.1
|
7, 8 |
1342 |
|
NR_133005.1 |
|
7, 8 |
1298 |
|
XM_011531588.2 |
XP_011529890.1
|
6, 7 |
1132 |
|
XM_011531589.1 |
XP_011529891.1
|
8, 9 |
1462 |
|
GenBank mRNA |
BC062476.1 |
- |
7, 8 |
1038 |
|
BC066227.1 |
- |
7, 8 |
1000 |
7002195 |
BC066228.1 |
- |
7, 8 |
1000 |
7002197 |
BC067419.1 |
- |
7, 8 |
1000 |
7002191 |
BC067420.1 |
- |
7, 8 |
1000 |
7002192 |
BC067421.1 |
- |
7, 8 |
1000 |
7002193 |
BC067422.1 |
- |
7, 8 |
1000 |
7002194 |
BC074771.2 |
- |
7, 8 |
1000 |
30915457 |
BC074786.2 |
- |
7, 8 |
999 |
30915344 |
CD014078.1 |
- |
7, 8 |
999 |
|
M12272.1 |
- |
7, 8 |
1052 |
|
X04299.1 |
- |
7, 8 |
1044 |
|
X04350.1 |
- |
5, 6 |
814 |
|
View Full Product Details