Gene Transcripts

Gene Symbol
FOLR1
Entrez Gene ID
2348
Gene Name
folate receptor 1
Gene Aliases
FBP, FOLR
Chromosome Location
Chr. 11: 72189558 - 72196323 on Build GRCh38
UniGene ID
Hs.73769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000802.3 NP_000793.1 3 261
NM_016724.2 NP_057936.1 4 512
NM_016725.2 NP_057937.1 3 446
NM_016729.2 NP_057941.1 2 281
GenBank mRNA AK223527.1 - 3 300
BC002947.2 - 3 393 3956659
BT007158.1 - 2 236
CR542019.1 - 2 236
J05013.1 - 3 447
KU257686.1 - 2 161
M28099.1 - 4 332
X62753.1 - 3 666

View Full Product Details