Gene Transcripts
- Gene Symbol
- PC
- Entrez Gene ID
- 5091
- Gene Name
- pyruvate carboxylase
- Gene Aliases
- PCB
- Chromosome Location
- Chr. 11: 66848522 - 66958418 on Build GRCh38
- UniGene ID
- Hs.89890
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000920.3 |
NP_000911.2
|
4, 5 |
414 |
|
NM_001040716.1 |
NP_001035806.1
|
5, 6 |
602 |
|
NM_022172.2 |
NP_071504.2
|
3, 4 |
369 |
|
XM_005274031.4 |
XP_005274088.1
|
3, 4 |
461 |
|
XM_005274032.4 |
XP_005274089.1
|
3, 4 |
343 |
|
XM_006718578.3 |
XP_006718641.1
|
3, 4 |
414 |
|
XM_011545086.2 |
XP_011543388.1
|
3, 4 |
730 |
|
XM_017017868.1 |
XP_016873357.1
|
6, 7 |
901 |
|
XM_017017869.1 |
XP_016873358.1
|
6, 7 |
772 |
|
XM_017017870.1 |
XP_016873359.1
|
5, 6 |
713 |
|
XM_017017871.1 |
XP_016873360.1
|
5, 6 |
584 |
|
XM_017017872.1 |
XP_016873361.1
|
4, 5 |
966 |
|
GenBank mRNA |
AK297705.1 |
- |
4, 5 |
414 |
|
BC011617.2 |
- |
4, 5 |
403 |
3946441 |
CN267865.1 |
- |
5, 6 |
607 |
|
DA542083.1 |
- |
4, 5 |
414 |
|
S72370.1 |
- |
3, 4 |
402 |
|
U04641.1 |
- |
3, 4 |
358 |
|
U30891.1 |
- |
4, 5 |
372 |
|
View Full Product Details