Gene Transcripts
- Gene Symbol
- Usp9y
- Entrez Gene ID
- 107868
- Gene Name
- ubiquitin specific peptidase 9, Y chromosome
- Gene Aliases
- Dffry, Fafl2
- Chromosome Location
- Chr. Y: 1298961 - 1459782 on Build GRCm38
- UniGene ID
- Mm.347376
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_148943.2 |
NP_683745.2
|
42 |
7329 |
|
GenBank mRNA |
AJ307017.1 |
- |
42 |
7329 |
|
View Full Product Details